Order Entry
Ireland
ContactUsLinkComponent
Anti-HSP101 Rabbit Polyclonal Antibody
Anti-HSP101 Rabbit Polyclonal Antibody
Catalog # AGRIAS08287
Supplier:  Agrisera
Anti-HSP101 Rabbit Polyclonal Antibody
Catalog # AGRIAS08287
Supplier:  Agrisera

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    HSP101
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    A. tequilana,
    A. thaliana,
    Thellungiella salsuginea
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Cross adsorption:
    No
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ambient
  • Immunogen:
    recombinant protein of Arabidopsis thaliana Hsp101/ClpB, C-terminal 145 amino acids P42730, At1g74310
  • Size:
    200 µl
  • Pk:
    200 µl

Specifications

About this item

Type: Primary
Antigen: HSP101
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: A. tequilana, A. thaliana, Thellungiella salsuginea