Anti-Deg1 Rabbit Polyclonal Antibody
Catalog # AGRIAS07246
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:DEG1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:A. thaliana
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:recombinant Deg1 of Arabidopsis thaliana O22609, At3g27925, overexpressed in E.coli
- Size:360 µg
- Pk:360 µG
Specifications
About this item
Type: Primary
Antigen: DEG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana