Anti-PEPCK Rabbit Polyclonal Antibody
Catalog # AGRIAS07241
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:PEPCK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Miscanthus giganteus,Panicum virgatum,Spartina patens,Zea mays,A. comosus,Phaseolus vulgaris,Mouse,Spartina alterniflora
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide well conserved in both PEPCK1 and 2 sequences from different plant species including Zea mays Q9SLZ0
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: PEPCK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. comosus, Mouse, Miscanthus giganteus, Panicum virgatum, Phaseolus vulgaris, Spartina alterniflora, Spartina patens, Zea mays