Anti-CCS Rabbit Polyclonal Antibody
Catalog # AGRIAS07219
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:CCS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Chlamydomonas reinhardtii
- Western blot:Yes
- Epitope:full length
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:The antibody was raised to the Arabidopsis thaliana full length CCs protein Q4ZJI5, fused to His-tag. Overexpressed using pET28a vector in Bl21 E. coli (Cd+) and purified using His-column. After purification, the His tag was cleaved using thrombin and then theprotein was purified using Resource-Q column connected to HPLC.
- Purification:purified using His-column
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: CCS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: full length
Host: Rabbit
Isotype:
Reactivity: Chlamydomonas reinhardtii