Anti-AtpB Rabbit Polyclonal Antibody
Catalog # AGRIAS05085
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen name:ATP Synthase beta subunit
- Antigen symbol:ATPB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:C. reinhardtii,A. thaliana
- Western blot:Yes
- Cross adsorption:No
- UniProtKB:P06541 and A8IQU3
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide derived from available plant, algal (chloroplastic and mitochondrial) and bacterial sequences of beta subunits of F-type ATP synthases, including Arabidopsis thaliana chloroplastic ATP synthase subunit beta
- Tested applications:Blue Native PAGE
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: ATPB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, C. reinhardtii