Anti-GOLM1 Mouse Monoclonal Antibody [clone: 2F3]
Catalog # ORIGTA600480
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:GOLM1
- Clonality:Monoclonal
- Clone:2F3
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Epitope:Full length
- Cross adsorption:No
- Form:Liquid
- Storage buffer:Stored in PBS (pH 7,4) containing 0,05% sodium azide and up to 5% trehalose
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human GOLM1 (NM_016548) produced in HEK293T cell.
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: 1:100-1:1000
Type: Primary
Antigen: GOLM1
Clonality: Monoclonal
Clone: 2F3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human