Order Entry
Ireland
ContactUsLinkComponent
Anti-Skor1 Rabbit Polyclonal Antibody
Anti-Skor1 Rabbit Polyclonal Antibody
Catalog # ORIGTA329597
Supplier:  OriGene
Anti-Skor1 Rabbit Polyclonal Antibody
Catalog # ORIGTA329597
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    SKI family transcriptional corepressor 1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Storage buffer:
    Lyophilized powder. Add 50 ul of distilled water. Final anti-Lbxcor1 antibody concentration is 1 mg/ml in PBS buffer with 2% sucrose. For longer periods of storage, store at -20C. Avoid repeat freeze-thaw cycles.
  • Molecular weight:
    100 kDa
  • Sequence:
    EPDKEDNHSTTADDLETRKSFSDQRSVSQPSPANTDRGEDGLTLDVTGTQ
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    The immunogen for anti-Lbxcor1 antibody: synthetic peptide directed towards the middle region of mouse Lbxcor1. Synthetic peptide located within the following region: EPDKEDNHSTTADDLETRKSFSDQRSVSQPSPANTDRGEDGLTLDVTGTQ
  • Size:
    50 μg
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: Skor1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse