Anti-AOC3 Rabbit Polyclonal Antibody
Catalog # ORIGTA325044
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:amine oxidase, copper containing 3
- Antigen symbol:AOC3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Cross adsorption:No
- Form:Liquid
- Amino acid number:613 to 640
- Storage buffer:Purified polyclonal antibody supplied in PBS with 0,09% (W/V) sodium azide.
- Molecular weight:84.62 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:This AOC3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 613-640 amino acids from the Central region of human AOC3.
- Purification:purified by Protein A chromatography, followed by affinity purification
- Size:400 μl
- Pk:400 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:10-50; Flow Cytometry: 1:10-50
Type: Primary
Antigen: AOC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human