Anti-POU5F1 Rabbit Polyclonal Antibody
Catalog # ORIGTA324706
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:POU5F1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Amino acid number:110 to 141
- Storage buffer:Purified polyclonal antibody supplied in PBS with 0,09% (W/V) sodium azide.
- Molecular weight:38.57 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:This OCT4 (OCT3) antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 110-141 amino acids from human OCT4 (OCT3).
- Purification:purified by saturated ammonium sulfate precipitation followed by dialysis against PBS
- Size:400 μl
- Pk:400 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:10-50
Type: Primary
Antigen: POU5F1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human