Anti-GYPA Mouse Polyclonal Antibody [clone: HIR2 (also GA-R2)]
Catalog # ORIGTA320342
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Glycophorin A
- Antigen symbol:GYPA
- Clonality:Polyclonal
- Clone:HIR2 (also GA-R2)
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Form:Liquid
- Storage buffer:Aqueous buffer, 0,09% sodium azide, may contain carrier protein/stabilizer
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: GYPA
Clonality: Polyclonal
Clone: HIR2 (also GA-R2)
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human