Anti-KREMEN1 Rabbit Polyclonal Antibody
Catalog # ORIGTA320040
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:KREMEN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Storage buffer:KREMEN1 Antibody is supplied in PBS containing 0,02% sodium azide.
- Molecular weight:54 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Rabbit polyclonal KREMEN1 antibody was raised against an 18 amino acid peptide near the carboxy terminus of human KREMEN1.
- Purification:purified by affinity chromatography via peptide column
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml; IHC; Immunofluorescence
Type: Primary
Antigen: KREMEN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat