Anti-MAP2K1 Mouse Monoclonal Antibody [clone: 8H1]
Catalog # ORIGCF506035
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:MAP2K1
- Clonality:Monoclonal
- Clone:8H1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Monkey,Human,Dog,Rat,Mouse
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:43.3 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human MAP2K1(NP_002746) produced in HEK293T cell.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:200-4000; Immunofluorescence: 1:100
Type: Primary
Antigen: MAP2K1
Clonality: Monoclonal
Clone: 8H1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey, Mouse, Rat