Anti-HIST1H3D Rabbit Polyclonal Antibody
Catalog # ORIGTA318931
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:HIST1H3D
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:near C-terminus
- Cross adsorption:No
- Form:Liquid
- Storage buffer:100 µg (0,5 mg/ml) affinity purified rabbit anti-Histone H3 polyclonal antibody in phosphate buffered saline (PBS), pH 7,2, containing 30% glycerol, 0,5% BSA, 0,01% thimerosal.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Synthetic peptide near the C-terminus of human Histone H3
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 0.5-4 ?g/ml; Immunochemistry: 20?g/ml
Type: Primary
Antigen: HIST1H3D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat