Anti-HIST2H2BE Rabbit Polyclonal Antibody
Catalog # ORIGTA318930
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:HIST2H2BE
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Storage buffer:100 µg (0,5 mg/ml) peptide affinity purified rabbit anti-Histone H2B polyclonal antibody in phosphate buffered saline (PBS), pH 7,2, containing 50% glycerol, 1% BSA, 0,02% thimerosal.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Synthetic peptide corresponding to the N-terminus of human Histone H2B
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 0.5-4 ?g/ml; Immunoprecipitation: 20-30?g/ml
Type: Primary
Antigen: HIST2H2BE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat