Anti-Angptl4 Rabbit Polyclonal Antibody
Catalog # ORIGTA318914
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Angiopoietin-like 4
- Antigen symbol:ANGPTL4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:ANGPTL4
- Storage buffer:100 µg (0,5 mg/ml) affinity purified rabbit polyclonal antibody in phosphate-buffered saline (PBS) containing 30% glycerol, 0,5% BSA, and 0,01% thimerosal.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Synthetic peptide surrounding amino acid 84 of mouse FIAF
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 0.5-4?g/ml
Type: Primary
Antigen: Angptl4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat