

N,N-Dimethylacetamide 99.99-100.00%, PESTINORM® for headspace gas chromatography
85397.290
:
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Purity (GC) :min. 99.8%
- Identity (IR) :Complying
- Residue after evaporation :max. 0.0003%w/w
- Water (KF) :max. 0.05%w/w
- Color (APHA Pt-Co) :max. 10
- GC-HS (Each class 1 solvent) :max. 1µg/g
- GC-HS (Each class 2 solvent) :max. 10µg/g
- GC-HS (Each class 3 solvent) :max. 50µg/g
- GC-HS (DMF):max. 20µg/g
- Transmittance (350 nm):Min. 98 %
- Transmittance (400 nm):Min. 99 %
- Headspace test for O.V.I's:Passes test
:
:87.12 g/mol
:13,03254
:166,1 °C (1013 hPa)
:–20 °C
:0,937 g/cm³ (20 °C)
:70 °C
:Ambient
:204-826-4
:2810
:6.1,III
:Filtered through a 0.2 µm filter, packaged under nitrogen.
:01-2119459339-27
:GHS08
:GHS07
:127-19-5
:CH₃CON(CH₃)₂
:MFCD00008686