Racks, magnetic separation, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Acrylic, clear.
- Independent sleeves make liquid addition and removal easier with less accidental fluid loss
- Powerful magnets quickly secure beads
- Spring plunger on tube sleeves allows beads to be sequestered at various heights along the tube wall
- Simply turn tube for efficient bead mixing
These magnetic separation racks has sequestered magnetic beads, so that rinse solutions and affinity ligands such as antibodies, streptavidin, or proprietary reagents for coupling nucleic acids can be added or decanted in assays. The 96-well PCR plate model has 24 magnets that snugly hold the plate while attracting beads to the wall of the tubes, whereas flat bottom microplate model has magnets covering entire footprint to accept all standard micro-plates with any number of wells. (i.e.,) 6, 12, 24, 48, 96.
Frequently Bought Together