Order Entry
Ireland
ContactUsLinkComponent
 
Anti-GRIN2C Rabbit Polyclonal Antibody
Catalog # BSBTPB9374
Supplier:  Boster Bio
undefined
Anti-GRIN2C Rabbit Polyclonal Antibody
Catalog # BSBTPB9374
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Glutamate receptor, ionotropic, N-methyl D-aspartate 2C
  • Antigen symbol:
    GRIN2C
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Gene ID:
    2905
  • Antigen synonyms:
    NMDAR2C|GluN2C|NR2C
  • UniProtKB:
    Q14957
  • Storage temperature:
    At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
  • Immunogen:
    E.coli-derived human NMDAR2C recombinant protein (Position: Q43-Q242). Human NMDAR2C shares 93% and 92.5% amino acid (aa) sequence identity with mouse and rat NMDAR2C, respectively.
  • Purification:
    Immunogen affinity purified.
  • Size:
    100 µg/vial
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Glutamate receptor ionotropic, NMDA 2C(GRIN2C) detection. Tested with WB in Human;Rat.

NMDAR2C, known as GRIN2C, is mapped to 17q25.1. Glutamate [NMDA] receptor subunit epsilon-3 is a protein that in humans is encoded by the GRIN2C gene. NMDA receptors are found in the central nervous system, are permeable to cations and have an important role in physiological processes such as learning, memory, and synaptic development. The receptor is a tetramer of different subunits (typically heterodimer of subunit 1 with one or more of subunits 2A-D), forming a channel that is permeable to calcium, potassium, and sodium, and whose properties are determined by subunit composition. Alterations in the subunit composition of the receptor are associated with pathophysiological conditions such as Parkinson's disease, Alzheimer's disease, depression, and schizophrenia. Alternative splicing results in multiple transcript variants.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.

WB: The detection limit for NMDAR2C is approximately 0.1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: HKDC1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human