Order Entry
Ireland
ContactUsLinkComponent
Anti-IRF5 Rabbit Antibody
Anti-IRF5 Rabbit Antibody
Catalog # BSBTPA2208
Supplier:  Boster Bio
undefined
Anti-IRF5 Rabbit Antibody
Catalog # BSBTPA2208
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    IRF5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    SLEB10
  • Amino acid number:
    456 to 472
  • UniProtKB:
    Q13568
  • Sequence:
    EQFKELHHIWQSQQRLQ
  • Storage temperature:
    –20 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human IRF5 (456-472aa EQFKELHHIWQSQQRLQ), identical to the related rat sequence, and different from the related mouse sequence by two amino acids.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Interferon regulatory factor 5(IRF5) detection. Tested with WB in Human;Mouse;Rat.

Interferon regulatory factor 5, also called IRF5 or SLEB10, is a protein that in humans is encoded by the IRF5 gene. IRF5 gene is mapped to 7q32.1. This gene encodes a member of the interferon regulatory factor(IRF) family, a group of transcription factors with diverse roles, including virus-mediated activation of interferon, and modulation of cell growth, differentiation, apoptosis, and immune system activity. Members of the IRF family are characterized by a conserved N-terminal DNA-binding domain containing tryptophan(W) repeats. Multiple transcript variants encoding different isoforms have been found for this gene, and a 30-nt indel polymorphism(SNP rs60344245) can result in loss of a 10-aa segment. This gene is a transcription factor involved in the induction of interferons IFNA and INFB and inflammatory cytokines upon virus infection.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.

Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: IRF5
Clonality:
Clone:
Conjugation:
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat