Order Entry
Ireland
ContactUsLinkComponent
Anti-Securin Rabbit Antibody
Anti-Securin Rabbit Antibody
Catalog # BSBTPA2240
Supplier:  Boster Bio
undefined
Anti-Securin Rabbit Antibody
Catalog # BSBTPA2240
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Securin
  • Antigen symbol:
    Securin
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    near the C-terminus
  • Cross adsorption:
    No
  • Form:
    Lyophilised
  • Amino acid number:
    146 - 161
  • Sequence:
    LMILDEERELEKLFQL
  • Storage temperature:
    –20 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human Securin (146-161aa LMILDEERELEKLFQL), different from the related rat sequence by four amino acids, and from the related mouse sequence by five amino acids.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Securin(PTTG1) detection. Tested with WB in Human.

Reconstitute with distilled water

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Securin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human