Anti-Adiponectin Rabbit Antibody
Catalog # BSBTPA2014-1
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Adipocyte Plasma Membrane Associated Protein
- Antigen symbol:APMAP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:near the C-terminus
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:a|Adipose most abundant gene transcript 1|30 kDa adipocyte complement-related protein|Adipocyte|ACRP30|C1q and collagen domain-containing protein
- Amino acid number:224 - 244
- Sequence:LYADNDNDSTFTGFLLYHDTN
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human ADIPOQ (224-244aa LYADNDNDSTFTGFLLYHDTN), different from the related mouse and rat sequences by one amino acid.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Adiponectin(ADIPOQ) detection. Tested with WB, IHC-P, IHC-F, ELISA in Human, Mouse, Rat.
Reconstitute with distilled water
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: APMAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat