Order Entry
Ireland
ContactUsLinkComponent
Weighing bottles, glass, KIMBLE®
Weighing bottles, glass, KIMBLE®
Catalog # KIML15157-2024
Supplier:  KIMBLEKONTES
undefined
Weighing bottles, glass, KIMBLE®
Catalog # KIML15157-2024
Supplier:  KIMBLEKONTES

Specifications

  • Ø×H:
    20×24 mm
  • NS:
    24/12
  • Capacity:
    35 ml
  • Pk:
    12

Specifications

About this item

Manufactured from 33 expansion, low extractable borosilicate.

  • Excellent temperature resistance
  • Excellent chemical resistance

These weighing bottles are designed for weighing small samples of coal.The cap-style closure fits over externally ground body.

Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements