Volumetric flasks, class A, cassia style, KIMBLE®
KIML28066-110
- NS:13
- Capacity:110 ml
- Height:260 mm
- Pk:2
- Tolerance:0,08 ± ml
Manufactured from 33 expansion, low extractable borosilicate glass, KIMAX®
- Calibrated to contain
- With a marking spot and a durable white ceramic enamel scale
This flask is used in the assay of oils of cinnamon, chenopodium, cloves, spearmint and other essential oils.
Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.