Chemical resistant overalls, 4570
: 3M
These disposable overalls made of PP with PE film are designed to help protect against hazardous dusts (Type 5), light liquid splashes (Type 6), low pressure liquid sprays (Type 4), and high pressure liquid jets (Type 3). They are also certified to offer protection against radioactive particulates and infective agents.
- Elasticated hood with three panel design
- Elasticated waist, wrists and ankles for convenience and freedom of movement
- Two-way zipper with sealable storm flap and chin flap
- Seams are taped with multi-layer clear tape
- Anti-static treated (inside only)
Ideal for use in chemical industries, e.g. for decontamination, tank cleaning, chemical mixing and handling, and pesticide spraying.
Colour: Grey
: Type 3/4/5/6, EN 1073-2, EN 14126, EN 1149-5; Cat. III.
- Catalog No:
- MMMA45703XL
- MMMA45704XL
- MMMA45702XL
- MMMA4570S
- MMMA4570L
- MMMA4570XL
- MMMA4570M
- Model:
- 4570
- 4570
- 4570
- 4570
- 4570
- 4570
- 4570
- Anti-static:
- +
- +
- +
- +
- +
- +
- +
- Garment application:
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
Frequently Bought Together