Order Entry
Ireland
ContactUsLinkComponent
Horizontal electrophoresis system, multiSUB choice
Horizontal electrophoresis system, multiSUB choice
Supplier:  CLEAVER SCIENTIFIC
Certificates
undefined
Horizontal electrophoresis system, multiSUB choice
Supplier:  CLEAVER SCIENTIFIC
Specifications
  • Catalog No:
  • CLEVMSCHOICE7
  • CLEVMSCHOICE10
  • CLEVMSCHOICETRIO
  • CLEVMSCHOICE15
  • Model:
  • multiSUB Choice 7
  • multiSUB Choice 10
  • multiSUB Choice TRIO
  • multiSUB Choice 15
  • Gel size W×L:
  • 150×70 mm
  • 150×100 mm
  • 150×150 mm
  • Sample capacity:
  • 70 samples
  • 140 samples
  • 210 samples
  • Buffer volume:
  • ~1000 ml
  • ~1000 ml
  • ~1000 ml
  • ~1000 ml
Specifications
Combs
Product Information
Frequently Bought Together