Order Entry
Ireland
ContactUsLinkComponent
Biological safety cabinets, class II, type A2, Purifier® Cell Logic®+
Biological safety cabinets, class II, type A2, Purifier® Cell Logic®+
Catalog # LBCP322691150
Supplier:  LABCONCO
   Energy Efficient
   Sustainable Materials
undefined
Biological safety cabinets, class II, type A2, Purifier® Cell Logic®+
Catalog # LBCP322691150
Supplier:  LABCONCO
   Energy Efficient
   Sustainable Materials

Specifications

  • Description:
    Biosafety cabinet, Purifier® Cell Logic®+, Class II, Type A2, nominal width 6 feet, UK plug
  • Seq:
    03
  • Environmentally Preferable:
    Energy Efficient,
    Sustainable Materials
  • Model:
    6' Purifier® Cell Logic®+
  • Electrical connection:
    230 V, 50/60 Hz, 8 A
  • Exhaust air volume:
    769,7 - 846,1 m³/h
  • Ext. W×D×H:
    1989×792×2268/2420 mm (with sash open)
  • Filters:
    HEPA filters, with Constant Airflow Profile™ (CAP) technology
  • Weight:
    410,5 kg
  • Pk:
    1

Specifications

About this item

These Biosafety cabinets provide personnel, product and environmental protection from hazardous particulates such as agents that require Biosafety Level 1, 2 or 3 containment. Other appropriate applications include work with antineoplastic drugs, genetic material, asbestos and additional substances that generate hazardous airborne particulates. All models are NSF compliant. Cabinets are designed to discharge HEPA-filtered exhaust air directly into the laboratory, or into an exhaust system through the optional canopy connection. When canopy-connected, the biosafety cabinets may be used for applications involving minute quantities of volatile toxic chemicals and tracer amounts of radionuclides as an adjunct to microbiological research.

  • Excels in user comfort and functionality
  • Ergonomic design: Inclination™ Technology Suite that includes the following: 10° angled sash, patented curved air grille with built in armrest, fully accessible controls and eye-level mounted colour display with MyLogic™ operating system
  • Provides intuitive programming, colour coded menus, status messages and alarms with diagnostic troubleshooting