Anti-Xenin 25 Rabbit Polyclonal Antibody
Catalog # USBIX1013-01
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Xenin 25
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Format:Serum
- Form:lyophilized powder
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Immunochemistry: 1:500
Type: Primary
Antigen: Xenin 25
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat