Anti-Trypsin Inhibitor, Ovomucoid Rabbit Polyclonal Antibody
Catalog # USBIT8682-05
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Trypsin Inhibitor, Ovomucoid
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Chicken
- Western blot:Yes
- Format:Serum
- Form:lyophilized powder
- Storage buffer:0,05% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000-1:5000; ELISA: 1:10000-1:50000
Type: Primary
Antigen: Trypsin Inhibitor, Ovomucoid
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken