Anti-Telomeric Repeat Binding Factor 2 Interacting Protein 1 Rabbit Monoclonal Antibody [clone: 11C1080]
USBIT2400-20C
- Antibody type:Primary
- Antigen name:Telomeric Repeat Binding Factor 2 Interacting Protein 1
- Antigen symbol:TERF2IP1
- Clonality:Monoclonal
- Clone:11C1080
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:10mMsodium HEPES, pH 7,5, 150mM sodium chloride, 100ug/ml BSA, less than 0,02% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 µl
- Pk:100 µl
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:100
Type: Primary
Antigen: Telomeric Repeat Binding Factor 2 Interacting Protein 1
Clonality: monoclonal
Clone: 11C1080
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey, Mouse, Rat