Order Entry
Ireland
ContactUsLinkComponent
Anti-STEAP4 Mouse Monoclonal Antibody [clone: 8H178]
Anti-STEAP4 Mouse Monoclonal Antibody [clone: 8H178]
Catalog # USBIS7500-02
Supplier:  US Biological
Anti-STEAP4 Mouse Monoclonal Antibody [clone: 8H178]
Catalog # USBIS7500-02
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    STEAP family member 4
  • Antigen symbol:
    STEAP4
  • Clonality:
    Monoclonal
  • Clone:
    8H178
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Form:
    lyophilized powder
  • Amino acid number:
    1 to 459
  • Storage buffer:
    PBS, 5% trehalose.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml (after reconstitution)
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: STEAP4 (STEAP family member 4)
Clonality: Monoclonal
Clone: 8H178
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human