Anti-S100 Calcium Binding Protein Rabbit Polyclonal Antibody
USBIS0052-05A
- Antibody type:Primary
- Antigen name:S100 Calcium Binding Protein
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Form:liquid
- Storage buffer:10mM PBS, pH 7,4, BSA, sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:250 µl
- Pk:250 µl
Recommended dilution: Immunochemistry: 1:50
Type: Primary
Antigen: S100 Calcium Binding Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human