Anti-Ras, pan, N,H,K Rabbit Monoclonal Antibody [clone: 7H56]
USBIR1198-01D
- Antibody type:Primary
- Antigen symbol:Ras, pan, N,H,K
- Clonality:Monoclonal
- Clone:7H56
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Yeast,Drosophila,Monkey,Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Format:Supernatant
- Form:liquid
- Storage buffer:10mMsodium HEPES, pH 7,5, 150mM sodium chloride, 0,1mg/ml BSA, <0,02% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: Ras, pan, N,H,K
Clonality: monoclonal
Clone: 7H56
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Drosophila, Human, Monkey, Mouse, Rat, Yeast