Specifications
- Antibody type:Primary
- Antigen symbol:PRG-1
- Clonality:Monoclonal
- Clone:8H159
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:lyophilized powder
- Storage buffer:PBS5% trehalose.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 0.5-1 ?g/ml; Western Blot: 1-2?g/ml
Type: Primary
Antigen: PRG-1
Clonality: monoclonal
Clone: 8H159
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human