Anti-Pregnancy-specific beta-1-glycoprotein Rabbit Polyclonal Antibody
Catalog # USBIP6000-93A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Pregnancy-specific beta-1-glycoprotein
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Recommended dilution: ELISA: 1:500-1:2000
Type: Primary
Antigen: Pregnancy-specific beta-1-glycoprotein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human