Anti-Platelet Derived Growth Factor Receptor, Type B Rabbit Monoclonal Antibody [clone: 5i127]
USBIP4258-20B
- Antibody type:Primary
- Antigen name:Platelet Derived Growth Factor Receptor, Type B
- Clonality:Monoclonal
- Clone:5i127
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat
- Western blot:Yes
- Form:liquid
- Modification:phospho-Tyr740
- Storage buffer:10mMsodium HEPES, pH 7,5, 150mM NaCl, 100ug/ml BSA, 50% glycerol.
- Storage temperature:Cool
- Concentration:100 ul (10 Western mini-blots)
- Shipping temperature:Ice
- Purification:purified
- Size:100 µl
- Pk:100 µl
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: Platelet Derived Growth Factor Receptor, Type B
Clonality: monoclonal
Clone: 5i127
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat