Anti-Phosphatidyl-4-phosphate-5-kinase, gamma Rabbit Polyclonal Antibody
Catalog # USBIP4071-76
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Phosphatidyl-4-phosphate-5-kinase, gamma
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 1mg/ml BSA, 0,1% sodium azidebefore the addition of 40% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 1:50000
Type: Primary
Antigen: Phosphatidyl-4-phosphate-5-kinase, gamma
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human