Anti-PEDF R Rat Monoclonal Antibody [clone: 11D53]
USBIP3140-04
- Antibody type:Primary
- Antigen symbol:PEDFR
- Clonality:Monoclonal
- Clone:11D53
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rat
- Isotype:IgG2a
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:lyophilized powder
- Amino acid number:162 to 253
- Storage buffer:PBS, 5% trehalose.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml (after reconstitition)
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Western Blot: 2?g/ml
Type: Primary
Antigen: PEDF R
Clonality: monoclonal
Clone: 11D53
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse