Anti-Nucleoside Diphosphate Kinase 6 Rabbit Polyclonal Antibody (FITC (Fluorescein))
USBIN7100-68N-FITC
- Antibody type:Primary
- Antigen name:Nucleoside Diphosphate Kinase 6
- Antigen symbol:NME6
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Form:liquid
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified by Protein G chromatography
- Size:200 µl
- Pk:200 µl
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!
Type: Primary
Antigen: Nucleoside Diphosphate Kinase 6
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human