Anti-NCSTN Mouse Monoclonal Antibody [clone: 10C300]
Catalog # USBIN2525-12
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Nicastrin
- Antigen symbol:NCSTN
- Clonality:Monoclonal
- Clone:10C300
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,1% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:500-1:1000
Type: Primary
Antigen: NCSTN (nicastrin)
Clonality: monoclonal
Clone: 10C300
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human