Order Entry
Ireland
ContactUsLinkComponent
Anti-Nerve Growth Factor Receptor, p75NTR - FITC Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 9G397]
Anti-Nerve Growth Factor Receptor, p75NTR - FITC Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 9G397]
Catalog # USBIN2050-40B
Supplier:  US Biological
Anti-Nerve Growth Factor Receptor, p75NTR - FITC Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 9G397]
Catalog # USBIN2050-40B
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Nerve Growth Factor Receptor, p75NTR - FITC
  • Clonality:
    Monoclonal
  • Clone:
    9G397
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Rat
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by affinity chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Recommended dilution: Immunofluorescence: 1:10-1:50; Flow Cytometry: 20 ?g/ml; ELISA: 1:5000 Immunochemistry: 1:10-1:50

Type: Primary
Antigen: Nerve Growth Factor Receptor, p75NTR - FITC
Clonality: monoclonal
Clone: 9G397
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat