Anti-SLC8A1 Mouse Monoclonal Antibody [clone: 9A71]
USBIN0560-21
- Antibody type:Primary
- Antigen name:solute carrier family 8 (sodium/calcium exchanger), member 1
- Antigen symbol:Solute carrier family 8, member 1
- Clonality:Monoclonal
- Clone:9A71
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgM
- Reactivity:Rabbit,Human,Guinea pig,Dog,Rat
- Western blot:Yes
- Format:ascites
- Form:liquid
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: SLC8A1 (solute carrier family 8 (sodium/calcium exchanger), member 1)
Clonality: monoclonal
Clone: 9A71
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Dog, Guinea pig, Human, Rabbit, Rat