Anti-Migration Inhibitory Factor Rabbit Polyclonal Antibody
Catalog # USBIM3895-07H
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Migration Inhibitory Factor
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:lyophilized powder
- Storage buffer:PBS, 5% BSA, 0,05% thimerosal, 0,05% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1-2?g/ml; Immunochemistry: 1-2?g/ml
Type: Primary
Antigen: Migration Inhibitory Factor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat