Order Entry
Ireland
ContactUsLinkComponent
 
Anti-MEK1, MEK2 Rabbit Monoclonal Antibody [clone: 7H39]
  USBIM2865-10H
Anti-MEK1, MEK2 Rabbit Monoclonal Antibody [clone: 7H39]
  USBIM2865-10H

 

  • Antibody type:
    Primary
  • Antigen symbol:
    MEK1/2
  • Clonality:
    Monoclonal
  • Clone:
    7H39
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Monkey,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Format:
    Supernatant
  • Form:
    liquid
  • Modification:
    phospho-Ser217, phospho-Ser221
  • Storage buffer:
    10mM HEPES, pH 7,5, 150mM sodium chloride,100ug/ml BSA, 50% glycerol and less than 0,02% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: MEK1, MEK2
Clonality: monoclonal
Clone: 7H39
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey, Mouse, Rat