Order Entry
Ireland
ContactUsLinkComponent
 
Anti-Matrix Metalloproteinase 1 Rabbit Monoclonal Antibody [clone: 9i45]
  USBIM2420-15B
Anti-Matrix Metalloproteinase 1 Rabbit Monoclonal Antibody [clone: 9i45]
  USBIM2420-15B

 

  • Antibody type:
    Primary
  • Antigen name:
    Matrix Metalloproteinase 1
  • Antigen symbol:
    MMP1
  • Clonality:
    Monoclonal
  • Clone:
    9i45
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Format:
    Supernatant
  • Form:
    liquid
  • Storage buffer:
    Tris-glycine, pH 7,4, 0,15M sodium chloride, 0,05% BSA, 0,01% sodium azide, 40% glycerol.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Recommended dilution: Flow Cytometry: 1:20; Western Blot: 1:1000; Immunochemistry: 1:50-250; Immunoprecipitation: 1:30

Type: Primary
Antigen: Matrix Metalloproteinase 1
Clonality: monoclonal
Clone: 9i45
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human