Anti-Matriptase Mouse Monoclonal Antibody [clone: 11C1095]
Catalog # USBIM2419-25B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Matriptase
- Clonality:Monoclonal
- Clone:11C1095
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:lyophilized powder
- Amino acid number:615 to 855
- Storage buffer:PBS, pH 7,4, 5% trehalose.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Matriptase
Clonality: monoclonal
Clone: 11C1095
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human