Anti-Lipid A, LPS Goat Polyclonal Antibody
Catalog # USBIL2496-20L
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Lipid A, LPS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- ImmunoFluorescence:Yes
- Isotype:IgG
- Form:liquid
- Storage buffer:PBS, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Type: Primary
Antigen: Lipid A, LPS
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: