Anti-Iron Responsive Element Binding Protein 2 Rabbit Polyclonal Antibody
Catalog # USBII8615-75B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Iron Responsive Element Binding Protein 2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Storage buffer:Tris-glycine and 150mM NaCl containing 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Iron Responsive Element Binding Protein 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat