Anti-IL34 Mouse Monoclonal Antibody (Biotin) [clone: 11D9]
Catalog # USBII8443-93E
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:interleukin 34
- Antigen symbol:IL34
- Clonality:Monoclonal
- Clone:11D9
- Conjugation:Biotin
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Environmentally Preferable:
- Epitope:N-terminal
- Form:lyophilized powder
- Storage buffer:PBS,BSA.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:250 µg
- Pk:250 µG
Specifications
About this item
Recommended dilution: ELISA: 0.5-2?g/ml
Type: Primary
Antigen: IL34 (interleukin 34)
Clonality: monoclonal
Clone: 11D9
Conjugation: Biotin
Epitope: N-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human