Order Entry
Ireland
ContactUsLinkComponent
Anti-NFKBIA Mouse Monoclonal Antibody [clone: 3A32]
Anti-NFKBIA Mouse Monoclonal Antibody [clone: 3A32]
Catalog # USBII2500-06A
Supplier:  US Biological
Anti-NFKBIA Mouse Monoclonal Antibody [clone: 3A32]
Catalog # USBII2500-06A
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, alpha
  • Antigen symbol:
    NFKBIA
  • Clonality:
    Monoclonal
  • Clone:
    3A32
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Monkey,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Format:
    ascites
  • Form:
    liquid
  • Modification:
    phospho-Ser32, phospho-Ser36
  • Storage buffer:
    10mM HEPES, pH 7,5, 150mM sodium chloride,0,1mg/ml BSA, <0,02% sodium azide, 50% glycerol.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: NFKBIA (nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, alpha)
Clonality: monoclonal
Clone: 3A32
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse, Rat