Anti-ECD Mouse Monoclonal Antibody [clone: 10D206]
USBIH7550-02
- Antibody type:Primary
- Antigen name:ecdysoneless homolog (Drosophila)
- Antigen symbol:ECD
- Clonality:Monoclonal
- Clone:10D206
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µl
- Pk:100 µl
Recommended dilution: ELISA: 1 ?g/ml-3ng/ml; Western Blot: 1:500-1:1000
Type: Primary
Antigen: ECD (ecdysoneless homolog (Drosophila))
Clonality: monoclonal
Clone: 10D206
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human